Learn More
CD14 Monoclonal Antibody (TuK4), PE-Texas Red, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD1417
Code : 50
Additional Details : Weight : 0.50000kg
Description
The R-phycoerythrin (PE)-Texas Red conjugate permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser.
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Monoclonal | |
PE-Texas Red | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Mouse | |
purified | |
ASR | |
929 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
TuK4 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
P08571 | |
Cd14 | |
Human CD14 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |