missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Mouse anti-Human, BUV661, Clone: MφP9 (also known as MφP-9), BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 741684
This item is not returnable.
View return policy
Description
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD14 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Hu CD14 BUV661 MφP9 50 μg | |
| Human Monocytes | |
| RUO | |
| CD14 | |
| Refrigerate (2 to 8°C) | |
| Human |
| Flow Cytometry | |
| MφP9 (also known as MφP-9) | |
| Brilliant Ultraviolet 661 | |
| Mouse | |
| 1 x 50 μg | |
| Primary | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction