Learn More
CD14 Monoclonal Antibody (TuK4), Pacific Orange™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD1430TR
Code : 50
Additional Details : Weight : 0.00800kg
Description
Our Pacific Orange™ dye is optimally excited by the violet laser, and it is recommended that a 575, 585 or 600 nm band pass filter be used for optimal detection. Pacific Blue™ and Pacific Orange™ dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm and 551 nm, respectively, facilitating two-color analysis.
CD14 Monoclonal antibody specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Monoclonal | |
Pacific Orange | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Mouse | |
purified | |
RUO | |
929 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
TuK4 | |
PBS with BSA and 0.1% sodium azide | |
P08571 | |
Cd14 | |
Human CD14 | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |