Learn More
CD138 (Syndecan-1) Rat, anti-Mouse, Unconjugated, Clone: 281-2, STEMCELL Technologies
Rat Monoclonal Antibody
Brand: Stemcell Technologies 60035.1
This item is not returnable.
View return policy
Description
- The 281-2 antibody reacts with an extracellular epitope on the core protein of mouse CD138 (Syndecan-1), a ∼60 - 100 kDa type 1 transmembrane glycoprotein expressed on the surface of pre-B cells, immature B cells, and normal and malignant plasma cells (but not mature circulating B cells)
- CD138 is also expressed on non-hematopoietic cells such as embryonic mesenchymal cells, endothelial, epithelial and neural cells
- CD138 is thought to act primarily as a receptor that modulates cell proliferation, cell migration and cell-matrix associations by linking the extracellular matrix to the cytoskeleton
- Theparin sulfate and chondroitin sulfate moieties attached to CD138 associate with several proteins, including collagens, fibronectin, tenascin, thrombospondin and certain cytokines
Specifications
| CD138 (Syndecan-1) | |
| Monoclonal | |
| Unconjugated | |
| 25 μg | |
| Mouse | |
| B Cells, Plasma |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 281-2 | |
| Rat | |
| Primary | |
| IgG2a κ |