Learn More
CD138 (Syndecan-1) Rat, anti-Mouse, APC, Clone: 281-2, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60035AZ
Encompass_Preferred
 View more versions of this product
Product Code. 17138671
  • £483.30 / 100µg
Description
  • The 281-2 antibody reacts with an extracellular epitope on the core protein of mouse CD138 (Syndecan-1), a ∼60 - 100 kDa type 1 transmembrane glycoprotein expressed on the surface of pre-B cells, immature B cells, and normal and malignant plasma cells (but not mature circulating B cells)
  • CD138 is also expressed on non-hematopoietic cells such as embryonic mesenchymal cells, endothelial, epithelial and neural cells
  • CD138 is thought to act primarily as a receptor that modulates cell proliferation, cell migration and cell-matrix associations by linking the extracellular matrix to the cytoskeleton
  • Theparin sulfate and chondroitin sulfate moieties attached to CD138 associate with several proteins, including collagens, fibronectin, tenascin, thrombospondin and certain cytokines
Specifications
Specifications
CD138 (Syndecan-1)
Monoclonal
APC
100 μg
Mouse
B Cells, Plasma
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
281-2
Rat
Primary
IgG2a κ