missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD138 (Syndecan-1) Mouse, anti-Human, PE, Clone: MI15, STEMCELL Technologies
Mouse Monoclonal Antibody
£194.40 - £480.60
Specifications
| Antigen | CD138 (Syndecan-1) |
|---|---|
| Clone | MI15 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17158341
|
Stemcell Technologies
60003PE.1 |
25 Tests |
£194.40
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17148341
|
Stemcell Technologies
60003PE |
100 Tests |
£480.60
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The MI15 antibody reacts with an extracellular epitope on CD138 (Syndecan-1), an ∼85 - 95 kDa type 1 transmembrane glycoprotein expressed on the surface of pre-B cells, immature B cells, and normal and malignant plasma cells (but not mature circulating B cells), as well as on non-hematopoietic cells such as embryonic mesenchymal cells, endothelial, epithelial and neural cells
- CD138 expression is used as a diagnostic marker for several types of tumors
- CD138 is thought to act primarily as a receptor that modulates cell proliferation, cell migration and cell-matrix associations by linking the extracellular matrix to the cytoskeleton
- Theparin sulfate and chondroitin sulfate moieties attached to CD138 associate with several proteins, including collagens, fibronectin, tenascin, thrombospondin and certain cytokines
- The MI15 antibody recognizes a different epitope than that of the clone DL-101 anti-CD138 antibody but blocks binding of clone B-B4
Specifications
| CD138 (Syndecan-1) | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| PE | |
| Cynomolgus, Human, Rhesus Monkey | |
| Primary |
| MI15 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title