missing translation for 'onlineSavingsMsg'
Learn More

CD138 (Syndecan-1) Monoclonal Antibody (DL-101), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen 14-1389-80

 View more versions of this product

Product Code. 15207227

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Description: The DL-101 monoclonal antibody reacts with human CD138, also known as syndecan-1. CD138 is a transmembrane protein containing chondroitin sulfate and heparin sulfate moieties responsible for binding to the extracellular matrix components. CD138 is not expressed by mature B cells but is present on pre-B cells and the finally differentiated plasma cells. Applications Reported: DL-101 has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections. Applications Tested: The DL-101 antibody has been tested by flow cytometric analysis of cell lines. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

CD138 (syndecan 1) is a transmembrane proteoglycan that can bind a variety of cytokines and modulate their activity, as well as the activity of extracellular matrix components and influence many developmental processes. CD138 is expressed mainly in differentiating keratinocytes and is transiently upregulated in all layers of the epidermis upon tissue injury. It is also highly expressed on plasma cells and can be detected even on fibroblasts, vascular smooth muscle cells and endothelial cells. Up-regulation and down-regulation of CD138 on the cell surface often correlates with the gain of cancerous characteristics. Serum levels of the shedded soluble sCD138 are used as a prognostic factor of cancerogenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD138 (Syndecan-1)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P18827
SDC1
Affinity chromatography
RUO
6382
4° C
Liquid
Flow Cytometry, Immunohistochemistry (Frozen)
DL-101
Unconjugated
SDC1
CD antigen 138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; SDC; SDC1; syndecan proteoglycan 1; syndecan-1; synstatin
Mouse
25 μg
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.