missing translation for 'onlineSavingsMsg'
Learn More

CD138 Mouse anti-Human, APC-R700, Clone: MI15  , BD Horizon™

Mouse Monoclonal Antibody

£132.86 - £367.22

Specifications

Antigen CD138
Clone MI15  
Applications Flow Cytometry
Classification Monoclonal
Conjugate APC-R700
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15804719
View Documents
BD Biosciences
566051
25 Tests
ONLINE EXCLUSIVE PRICE
£139.00
£132.86 / Each
Save £6.14
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15894709
View Documents
BD Biosciences
566050
100 Tests
ONLINE EXCLUSIVE PRICE
£382.00
£367.22 / Each
Save £14.78
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD138 Monoclonal antibody specifically detects CD138 in Human samples. It is validated for Flow cytometry.

CD138 Monoclonal antibody specifically detects CD138 in Human samples. It is validated for Flow cytometry.
Specifications

Specifications

CD138
Flow Cytometry
APC-R700
Mouse
Cell Biology
Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide.
Human U266 and XG-1 Myeloma Cell Lines
Primary
Vol. Per test: 5μL
MI15  
Monoclonal
Purified
RUO
Human
Syndecan; SDC; Syndecan 1; SDC1; SYND1
IgG1 κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.