missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD138 Monoclonal Antibody (D2), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550925
This item is not returnable.
View return policy
Description
CD138 (syndecan 1) is a transmembrane proteoglycan that can bind a variety of cytokines and modulate their activity, as well as the activity of extracellular matrix components and influence many developmental processes. CD138 is expressed mainly in differentiating keratinocytes and is transiently upregulated in all layers of the epidermis upon tissue injury. It is also highly expressed on plasma cells and can be detected even on fibroblasts, vascular smooth muscle cells and endothelial cells. Up-regulation and down-regulation of CD138 on the cell surface often correlates with the gain of cancerous characteristics. Serum levels of the shedded soluble sCD138 are used as a prognostic factor of cancerogenesis.
Specifications
| CD138 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| P18827, P26260 | |
| Sdc1 | |
| Recombinant Human SDC1 protein, Gln23-Leu253 (Accession P18827). | |
| 200 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a κ |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| D2 | |
| Unconjugated | |
| Sdc1 | |
| AA408134; AA409076; CD antigen 138; CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; HSPG; sCD138; SDC; Sdc1; soluble CD138; Sstn; syn-1; Synd; SYND1; Synd-1; SYNDECA; syndecan; syndecan 1; syndecan proteoglycan 1; Syndecan1; syndecan-1; synstatin | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 25216, 6382 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction