Learn More
CD137 Ligand Mouse anti-Human, Brilliant Violet 421, Clone: C65-485, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 744392
Description
The C65-485 monoclonal antibody specifically recognizes the type II membrane protein known as 4-1BB Ligand (4-1BB-L) or CD137 Ligand (CD137L). This molecule belongs to the TNF superfamily and is encoded by TNFSF9 /(Tumor necrosis factor superfamily member 9). CD137 Ligand has been reported to have costimulatory function on activated T cells. Reports suggest that stimulation of activated T cells via CD137 Ligand does not require the presence of other costimulatory molecules, including CD28. Some studies have reported human CD137 Ligand to be expressed preferentially on primary B cells and B-cell lines. The C65-485 antibody is able to react with recombinant human 4-1BB Ligand (rh4-1BB-L/CD137L) bound to CD137 (4-1BB receptor) expressed on phytohemagglutinin (PHA)-stimulated PBMC.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications
| CD137 Ligand | |
| C65-485 | |
| Brilliant Violet 421 | |
| 4-1BB Ligand, 4-1BB-L, TNFSF9, TNLG5A | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.