Learn More
CD137 Ligand (4-1BB Ligand) Monoclonal Antibody (TKS-1), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£72.75 - £219.00
Specifications
Antigen | CD137 Ligand (4-1BB Ligand) |
---|---|
Clone | TKS-1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15519316
|
Affymetrix eBioscience
46-5901-82 |
100 μg |
£219.00
100µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15509316
|
Affymetrix eBioscience
46-5901-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The TKS-1 monoclonal antibody reacts with mouse 4-1BB Ligand (4-1BBL), a type II transmembrane protein and a member of the tumor necrosis factor superfamily. 4-1BBL is expressed by activated antigen presenting cells including B cells and macrophages. Interaction of 4-1BBL with 4-1BB (CDw137) is reported to deliver a costimulatory signal important in the T-APC interaction. Applications Reported: This TKS-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TKS-1 antibody has been tested by flow cytometric analysis of stimulated mouse spleen cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD137 Ligand (4-1BB Ligand) Monoclonal antibody specifically detects CD137 Ligand (4-1BB Ligand) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD137 Ligand (4-1BB Ligand) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P41274 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
TKS-1 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Rat | |
Mouse | |
21950 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.