missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD137 (4-1BB) Monoclonal Antibody (4B4 (4B4-1)), APC-eFluor™ 780, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£179.00 - £345.00
Specifications
| Antigen | CD137 (4-1BB) |
|---|---|
| Clone | 4B4 (4B4-1) |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17123913
|
Invitrogen™
47-1379-41 |
25 Tests |
£179.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17133913
![]() |
Invitrogen™
47-1379-42 |
100 Tests |
£345.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD137, also known as TNFRSF9 or 4-1BB, is an inducible costimulatory molecule expressed mainly on activated T cells. Its ligand, known as 4-1BBL, is expressed on activated macrophages, mature B cells, hematopoietic stem cells, and myeloid progenitor cells. CD137 signaling leads to maintaining the survival of activated T cells and CD8+ memory T cells, and clonal expansion of T cells, but also to suppressing myelopoiesis and dendritic cell development. Triggered CD137 induces a cytokine release profile regulating peripheral monocyte survival. Soluble forms of CD137 may provide negative control mechanism for some immune responses.Specifications
| CD137 (4-1BB) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 4 1 BB; 41BB; 4-1BB; 4-1BB ligand receptor; A930040I11Rik; AA408498; AI325004; Cd137; CD137 antigen; CDw137; EGK_00183; homolog of mouse 4-1BB; ILA; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; Ly63; receptor protein 4-1BB; s4 1 BB; s4 1BB; s41BB; sCD137; secreted CD137 antigen; soluble 4 1 BB; soluble 4 1BB; soluble 41BB; soluble CD137; T cell antigen ILA; T-cell antigen 4-1BB; T-cell antigen 4-1BB homolog; T-cell antigen ILA; TNF receptor superfamily member 9; TNFRSF9; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9 | |
| TNFRSF9 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| TNFRSF9 |
| 4B4 (4B4-1) | |
| Flow Cytometry | |
| APC-eFluor 780 | |
| Mouse | |
| Human | |
| Q07011 | |
| 3604 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts