missing translation for 'onlineSavingsMsg'
Learn More

CD137 (4-1BB) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ CD137 (4-1BB) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

£103.00 - £601.80

Specifications

Antigen CD137 Ab-1
Clone BBK-2
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Form Concentrated
View More Specs

Products 8
Product Code Brand Conjugate Concentration Quantity Price Quantity & Availability  
Product Code Brand Conjugate Concentration Quantity Price Quantity & Availability  
12601817
View Documents Promotion Details
Epredia
MS-621-B0
Biotin 200μg/mL 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12611817
View Documents Promotion Details
Epredia
MS-621-B1
Biotin 200μg/mL 500 μL
£448.00
500µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12691807
View Documents Promotion Details
Epredia
MS-621-B
Biotin 200μg/mL 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12631817
View Documents Promotion Details
Epredia
MS-621-P0
Unconjugated 200μg/mL 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12641817
View Documents Promotion Details
Epredia
MS-621-P1
Unconjugated 200μg/mL 500 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12621817
View Documents Promotion Details
Epredia
MS-621-P
Unconjugated 200μg/mL 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12651817
View Documents Promotion Details
Epredia
MS-621-P1ABX
Unconjugated 1mg/mL 100 μL
£384.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12661817
View Documents Promotion Details
Epredia
MS-621-PABX
Unconjugated 1mg/mL 200 μL
£558.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-1BB (also known as CD137) is an inducible receptor-like protein expressed on the cell surface of activated splenic T cells and thymocytes. It exists as both a monomer and a dimer on the surface of activated T cells. 4-1BB is structurally related to the members of NGFR/TNFR superfamily which are characterized by the presence of three-six patterns of a cysteine-rich motif in their extracellular domains. Other members of this family include low affinity NGFR, two receptors for TNF (TNFR-I & TNFR-II), CD30, CD40, OX40, Fas, and CD27. These molecules are involved in cell growth, survival, and death processes. The cytoplasmic domain of 4-1BB include two runs of acidic amino acids, a potential p56lck binding site, five consecutive glycines at the C-terminus, and four potential phosphorylation sites: one tyrosine, two threonine, and one serine.

Host Species: Mouse

Clone: BBK-2

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Ectodomain

Immunogen: Ectodomain of human 4-1BB recombinant protein

Molecular Weight: 30kDa

Positive Control: Spleen, thymus or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD137 Ab-1
Flow Cytometry, Immunofluorescence
Concentrated
RUO
Human
IgG1 κ
BBK-2
Monoclonal
Mouse
Apoptosis
Ectodomain of human 4-1BB recombinant protein
Primary
Videos
SDS
Documents

Documents

Product Certifications