missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD135 (FLT3) Mouse anti-Human, BUV615, Clone: 4G8, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 752370
This item is not returnable.
View return policy
Description
CD135 (FLT3) Monoclonal antibody specifically detects CD135 (FLT3) in Human samples. It is validated for Flow Cytometry.
Specifications
| CD135 (FLT3) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Hu CD135 BUV615 4G8 50 μg | |
| Human FLT3 Transfected Cell Line | |
| RUO | |
| CD135 (FLT3) | |
| Refrigerate (2 to 8°C) | |
| Human |
| Flow Cytometry | |
| 4G8 | |
| Brilliant Ultraviolet 615 | |
| Mouse | |
| 1 x 50 μg | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction