missing translation for 'onlineSavingsMsg'
Learn More

CD135 (Flt3) Monoclonal Antibody (BV10A4H2), APC, eBioscience™, Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£89.25 - £241.00

Specifications

Antigen CD135 (Flt3)
Clone BV10A4H2
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15538006
View Documents
Invitrogen
17-1357-41
25 Tests This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15548006
GREENER_CHOICE
View Documents
Invitrogen™
17-1357-42
100 Tests
£241.00
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The BV10A4H2 monoclonal antibody reacts with human CD135, also known as Flk2/Flt3. A member of the tyrosine kinase receptor family, this 135-150 kDa molecule is expressed by multipotential progenitor cells including primitive B cell and myelomonocytic progenitors in fetal liver and adult bone marrow. Applications Reported: The BV10A4H2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This BV10A4H2 antibody has been pre-titrated and tested by flow cytometric analysis of REH cell line. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family. The receptor is activated by binding of the fms-related tyrosine kinase 3 ligand to the extracellular domain, which induces homodimer formation in the plasma membrane leading to autophosphorylation of the receptor. The activated receptor kinase subsequently phosphorylates and activates multiple cytoplasmic effector molecules in pathways involved in apoptosis, proliferation, and differentiation of hematopoietic cells in bone marrow. Mutations that result in the constitutive activation of this receptor result in acute myeloid leukemia and acute lymphoblastic leukemia.
Specifications

Specifications

CD135 (Flt3)
5 μL/Test
Monoclonal
Liquid
RUO
P36888
FLT3
IgG1
Affinity chromatography
Antibody
BV10A4H2
Flow Cytometry
APC
Mouse
Human
2322
BV-173 leukemic cell line
Primary
4° C, store in dark, DO NOT FREEZE!
FLT3
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.