Learn More
CD135 (Flt3) Monoclonal Antibody (A2F10), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£89.50 - £254.00
Specifications
Antigen | CD135 (Flt3) |
---|---|
Clone | A2F10 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15248669
|
Affymetrix eBioscience
12-1351-81 |
50 μg |
£89.50
50µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15258669
|
Affymetrix eBioscience
12-1351-82 |
100 μg |
£157.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15268669
|
Affymetrix eBioscience
12-1351-83 |
200 μg |
£254.00
200µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The A2F10 monoclonal antibody reacts with mouse CD135, also known as Flk2/Flt3. A member of the tyrosine kinase receptor family, this 135-150 kDa molecule is expressed by primitive progenitor cells in fetal liver and adult bone marrow. Two-color staining of adult mouse bone marrow cells with A2F10 and RA3-6B2 (CD45R/B220) or M1/70 (CD11b) reveals Flk-2-positive subpopulations of B cells or myeloid cells, respectively. Applications Reported: The A2F10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This A2F10 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family. The receptor is activated by binding of the fms-related tyrosine kinase 3 ligand to the extracellular domain, which induces homodimer formation in the plasma membrane leading to autophosphorylation of the receptor. The activated receptor kinase subsequently phosphorylates and activates multiple cytoplasmic effector molecules in pathways involved in apoptosis, proliferation, and differentiation of hematopoietic cells in bone marrow. Mutations that result in the constitutive activation of this receptor result in acute myeloid leukemia and acute lymphoblastic leukemia.Specifications
CD135 (Flt3) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
B230315G04; CD135; CD135 antigen; fetal liver kinase 2; Fetal liver kinase-2; FL cytokine receptor; FLK2; Flk-2; FLT3; Flt-3; fms related tyrosine kinase 3; FMS-like tyrosine kinase 3; fms-related tyrosine kinase 3; growth factor receptor tyrosine kinase type III; Ly72; OTTHUMP00000214829; Receptor-type tyrosine-protein kinase FLT3; RP11-153M24.3; stem cell tyrosine kinase 1; STK1; STK-1; tyrosine-protein kinase FLT3; tyrosine-protein kinase receptor flk-2; wmfl | |
FLT3 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
FLT3 |
A2F10 | |
Flow Cytometry | |
PE | |
Rat | |
Mouse | |
Q00342 | |
14255 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.