missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD133 Mouse anti-Human, BV421, Clone: 293C3, BD Horizon™
Mouse Monoclonal Antibody
£168.55 - £525.27
Specifications
| Antigen | CD133 |
|---|---|
| Clone | 293C3 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15845179
|
BD Biosciences
566598 |
25 μg |
£173.00 £168.55 / Each Save £4.45 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15815179
|
BD Biosciences
566595 |
0.1 mg |
£550.00 £525.27 / Each Save £24.73 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD133 Monoclonal antibody specifically detects CD133 in Human samples. It is validated for Flow Cytometry,Immunofluorescence,Immunohistochemistry (Frozen).Specifications
| CD133 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG2b κ | |
| Refrigerate (2 to 8°C) |
| 293C3 | |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Brilliant Violet 421 | |
| Mouse | |
| Human | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title