missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD133 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£240.00 - £353.00
Specifications
| Antigen | CD133 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18264774
|
Novus Biologicals
NBP2-59787 |
100 μL |
£353.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654127
|
Novus Biologicals
NBP2-59787-25ul |
25 μL |
£240.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD133 Polyclonal specifically detects CD133 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CD133 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 8842 | |
| This antibody was developed against a recombinant protein corresponding to the amino acid sequence:QYNTTKDKAFTDLNSINSVLGGGILDRLRPNIIPVLDEIKSMATAIKETKEALENMNSTLKSLHQQSTQLSSSLTSVKTSLRSSLNDPLCLVHPSSETCNSIRLSLS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Core ESC Like Genes, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1 | |
| PROM1 | |
| IgG | |
| Protein A purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title