missing translation for 'onlineSavingsMsg'
Learn More

CD133 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB120-16518JF646

424.15 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18730243

  • £532.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD133 Polyclonal antibody specifically detects CD133 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD133
Polyclonal
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat, Pig
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Janelia Fluor 646
AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1
Synthetic peptide corresponding to a C-terminal region of CD133 (within amino acids 750-865).
0.1 mL
Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Cellular Signaling, Core ESC Like Genes, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
8842
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.