Learn More
CD130 (GP130) Monoclonal Antibody (KGP130), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 12-1302-82
Code : Z1
Additional Details : Weight : 128.73000kg
Description
Description: The KGP130 monoclonal antibody reacts with mouse gp130, also known as CD130. gp130 is a 130 kD Type-I transmembrane glycoprotein containing a 597 amino acid extracellular domain, a single transmembrane domain, and a 277 amino acid cytoplasmic domain. gp130 is a subunit of several heterodimeric cell-surface receptors, including receptors for IL-6, IL-11, IL-27, Oncostatin M, and Leukemia Inhibitory Factor (LIF). The gp130 protein has also been found to exist in a soluble form, which is capable of inhibiting IL-6 signaling. gp130 is expressed mainly on T cells, monocytes, endothelial cells, activated B cells, and plasma cells, and is expressed at lower levels on most leukocytes and epithelial cells. In response to ligand binding, gp130 becomes tyrosine phosphorylated, leading to activation of several signaling pathways including the PI3 kinase, Ras-MAPk and Stat pathways. Preincubation of mouse splenic T cells with IL-6 is capable of blocking the interaction of KGP130 with gp130, suggesting that the KGP130 binding site lies near the IL-6 interaction domain. Reactivity of KGP130 towards human gp130 has not been observed. Applications Reported: This KGP130 antibody has been reported for use in flow cytometric analysis. Applications Tested: This KGP130 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test.
The protein encoded by this gene is a signal transducer shared by many cytokines, including interleukin 6 (IL6), ciliary neurotrophic factor (CNTF), leukemia inhibitory factor (LIF), and oncostatin M (OSM). This protein functions as a part of the cytokine receptor complex. The activation of this protein is dependent upon the binding of cytokines to their receptors. vIL6, a protein related to IL6 and encoded by the Kaposi sarcoma-associated herpesvirus, can bypass the interleukin 6 receptor (IL6R) and directly activate this protein. Knockout studies in mice suggested a critical role of the gene encoding this protein in regulating myocyte apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
CD130 (GP130) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q00560 | |
IL6ST | |
Affinity chromatography | |
RUO | |
16195 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
KGP130 | |
PE | |
IL6ST | |
5133400A03Rik; AA389424; Ac1055; BB405851; CD130; CD130 antigen; CDW130; D13Ertd699e; EGK_16499; Gp130; gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form; gp130, oncostatin M receptor; GP130-RAPS; IL-6; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6RB; IL-6R-beta; IL6R-beta; IL6ST; interleukin 6 signal transducer; interleukin 6 signal transducer (gp130, oncostatin M receptor); interleukin 6 signal transducer receptor; interleukin receptor beta chain; interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; interleukin-6 signal transducer receptor; membrane glycoprotein; membrane glycoprotein 130; membrane glycoprotein gp130; oncostatin-M receptor subunit alpha | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.