missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD127 Rat anti-Mouse, PerCP-Cy5.5, Clone: A7R34, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A14799
This item is not returnable.
View return policy
Description
Specifications
| CD127 | |
| Monoclonal | |
| PerCP-Cyanine5.5 | |
| Rat | |
| RUO | |
| Mouse | |
| Liquid |
| Flow Cytometry | |
| A7R34 | |
| PBS, pH 7.2, with 0.1% gelatin and 0.09% sodium azide | |
| 25 μg | |
| Primary | |
| Store at 2 - 8 °C. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction