Learn More
Invitrogen™ CD127 Monoclonal Antibody (A7R34), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ 12-1271-81
Description
Description: The A7R34 monoclonal antibody reacts with mouse CD127, the high affinity alpha subunit of the mouse IL-7 receptor. IL-7 receptor alpha chain is expressed by immature B cells in the bone marrow, double-negative (CD4-CD8-), single-positive (CD4+ and CD8+), but not double-positive (CD4+CD8+) thymocytes. In the periphery, mature T cells express CD127 at low level. A7R34 inhibits binding of IL-7 to its receptor and has been used in in vivo and in vitro studies to elucidate the role of IL-7 in T and B cell development and activation. Binding of A7R34 blocks the binding of SB/199, another antibody which recognizes mouse CD127. Applications Reported: The A7R34 antibody has been reported for use in flow cytometric analysis. Applications Tested: The A7R34 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD127, also known as the Interleukin-7 receptor alpha chain (IL-7Ralpha), is a type I glycoprotein with a molecular weight of 75-80 kDa. It forms a multi-functional receptor complex with CD132, the common gamma chain, to create the IL-7 receptor (IL-7R), which is crucial for lymphopoiesis. The active IL-7 receptor is an alpha/gamma chain heterodimer, where the gamma chain also associates with the interleukin-2 receptor and primarily activates signal transduction, while the alpha chain determines specific signaling events through its association with cytoplasmic signaling molecules. CD127 is expressed by immature B cells through the early pre-B stage, thymocytes during various stages of development, and most mature T cells, with transient down-regulation upon activation. It promotes the proliferation of precursor B cells, thymocytes, T cell progenitors, and mature CD4+ and CD8+ T cells. Binding of IL-7 to CD127 results in signal transduction through several tyrosine kinase pathways, including the Jak/STAT pathway, and is indispensable for lymphocyte development and the control of homeostatic proliferation of T cells in the periphery. Additionally, IL-7R signaling is involved in the regulation of T cell receptor (TCR) locus rearrangement in gamma delta T cells. Interestingly, CD127 expression is down-regulated on CD4+CD25+ regulatory T cells (T regs). While CD4 and CD25 co-expression is commonly used to identify T regs, this method may include cells without suppressive activity. CD4+CD25+ cells that have down-regulated CD127 expression are significantly enriched for the regulatory T cell population, as defined by the expression of the T reg-specific transcription factor Foxp3 and their suppressive activity in vitro. Diseases associated with CD127 dysfunction include severe combined immunodeficiency and T cell negative/B cell negative/NK positive severe combined immunodeficiency.
Specifications
| CD127 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P16872 | |
| Il7r | |
| Affinity chromatography | |
| RUO | |
| 16197 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| A7R34 | |
| PE | |
| Il7r | |
| CD127; CD127 antigen; CDW127; IL 7 receptor; IL 7R a; IL 7R subunit alpha; IL 7R α; IL 7Ra; IL 7Rα; IL7 receptor; IL-7 receptor alpha chain; IL7 receptor subunit alpha; IL-7 receptor subunit alpha; IL7R; IL-7R; IL7R a; IL7R alpha; IL7R subunit alpha; IL-7R subunit alpha; IL7R α; IL7RA; IL-7RA; IL-7Ralpha; IL-7R-alpha; IL7Rα; ILRA; interleukin 7 receptor; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin7 receptor subunit alpha; interleukin-7 receptor subunit alpha; MGC107557; sCD127; soluble IL 7 receptor; soluble IL 7R | |
| Rat | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.