missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD127 Monoclonal Antibody (eBioRDR5), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£247.00
Specifications
Antigen | CD127 |
---|---|
Clone | eBioRDR5 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | eFluor 660 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15529906
|
Affymetrix eBioscience
50-1278-42 |
100 Tests |
£247.00
100 tests |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Interleukin-7 is a glycoptorein involved in the regulation of lymphopoiesis. Response of cells to IL7 is dependent on the presence of the interleukin 7 receptor (IL7R); the active receptor is a alpha/gamma chain heterodimer. The gamma(c) chain, which also associates with the interleukin-2 receptor, serves primarily to activate signal transduction by the IL7R complex, while the alpha chain of IL7R determines specific signaling events through its association with cytoplasmic signaling molecules.
CD127 Monoclonal antibody specifically detects CD127 in Human samples. It is validated for Flow Cytometry.Specifications
CD127 | |
Flow Cytometry | |
eFluor 660 | |
Mouse | |
Human | |
3575 | |
Primary | |
Antibody |
eBioRDR5 | |
Monoclonal | |
Liquid | |
RUO | |
P16871 | |
IgG1 κ | |
Affinity chromatography |
For Research Use Only.