missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD127 Monoclonal Antibody (A7R34), Brilliant Ultra Violet™ 661, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£185.00 - £374.00
Specifications
| Antigen | CD127 |
|---|---|
| Clone | A7R34 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17835842
|
Invitrogen™
376-1271-80 |
25 μg |
£185.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17815932
![]() |
Invitrogen™
376-1271-82 |
100 μg |
£374.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD127 (Interleukin-7, IL-7) is a glycoprotein involved in the regulation of lymphopoiesis. The response of cells to CD127 is dependent on the presence of the interleukin 7 receptor (IL7R); the active receptor is an alpha/gamma chain heterodimer. CD127 consists of an alpha chain and a gamma chain. The gamma(c) chain, which also associates with the interleukin-2 receptor, serves primarily to activate signal transduction by the IL7R complex, while the alpha chain of IL7R determines specific signaling events through its association with cytoplasmic signaling molecules. CD127 promotes the proliferation of precursor B cells, thymocytes, T cell progenitors, and mature CD4+ and CD8+ T cells. The biological effects of IL7 are mediated by the binding of IL7 to the specific cell surface receptor. Diseases associated with CD127 dysfunction include severe combined immunodeficiency and T cell negative/B cell negative/NK positive severe combined immunodeficiency.Specifications
| CD127 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| CD127; CD127 antigen; CDW127; IL 7 receptor; IL 7R a; IL 7R subunit alpha; IL 7R α; IL 7Ra; IL 7Rα; IL7 receptor; IL-7 receptor alpha chain; IL7 receptor subunit alpha; IL-7 receptor subunit alpha; IL7R; IL-7R; IL7R a; IL7R alpha; IL7R subunit alpha; IL-7R subunit alpha; IL7R α; IL7RA; IL-7RA; IL-7Ralpha; IL-7R-alpha; IL7Rα; ILRA; interleukin 7 receptor; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin7 receptor subunit alpha; interleukin-7 receptor subunit alpha; MGC107557; sCD127; soluble IL 7 receptor; soluble IL 7R | |
| Il7r | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| A7R34 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 661 | |
| Rat | |
| Mouse | |
| P16872 | |
| 16197 | |
| IgG2a κ | |
| Affinity chromatography | |
| Il7r |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title