missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD127 / IL-7Ra Rat, V450, Clone: A7R34, enQuireBio™
Rat Monoclonal Antibody
£155.00 - £320.00
Specifications
Antigen | CD127 / IL-7Ra |
---|---|
Clone | A7R34 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952208
|
EnquireBio
QAB55-V450-25UG |
25 μg |
£155.00
25µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942208
|
EnquireBio
QAB55-V450-100UG |
100 μg |
£320.00
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD127 / IL-7Ra | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
16172 | |
IgG2a κ | |
Protein A/G Purified |
A7R34 | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
NP_032385 | |
Il17ra | |
Primary | |
Store at 2-4°C. |