Learn More
CD127 / IL-7Ra Rat, FITC, Clone: A7R34, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB55-F-500ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15962198
  • £265.00 / 500µg
Estimated Shipment: 04-07-2024
to see stock.
Specifications
CD127 / IL-7Ra
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Il17ra
Protein A/G Purified
RUO
16172
IgG2a κ
Flow Cytometry
A7R34
FITC
NP_032385
Rat
500 μg
Primary
Store at 2-4°C.