Learn More
CD123 Mouse anti-Human, PE, Clone: 7G3, BD
Mouse Monoclonal Antibody
£118.71 - £296.78
Specifications
| Antigen | CD123 |
|---|---|
| Clone | 7G3 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15860729
|
BD Biosciences
561058 |
25 μg |
£121.00 £118.71 / Each Save £2.29 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15847758
|
BD Biosciences
554529 |
0.2 mg |
£301.00 £296.78 / Each Save £4.22 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 7G3 monoclonal antibody specifically reacts with human CD123, the 70 kDa IL-3 Receptor α (IL-3Rα) chain. CD123 associates with CD131, the 120-140 kDa Common β chain to form the IL-3 Receptor Complex. CD131 is shared with the receptors for interleukins IL-5 and GM-CSF. IL-3Rα is expressed on hematopoietic progenitors and plays an important role in hematopoietic progenitor cell growth and differentiation. It is also expressed by mast cells, macrophages and a CD5+ B cell subset. This antibody has been reported to block the binding of 125I-IL-3 to high and low affinity IL-3 receptors. In functional experiments, this antibody was found to inhibit acute myeloid leukemia cell proliferation, basophil histamine release, endothelial cell-mediated IL-8 secretion, and neutrophil transmigration. This antibody has been reported to be useful for immunoprecipitation, Western blot and immunofluorescent staining for flow cytometry. At the Fifth HLDA Workshop, the human IL-3 receptor was designated CD123.
Specifications
| CD123 | |
| 0.2mg/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human IL-3Ra-transfected cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 7G3 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified | |
| IL3RA; IL-3RA; IL-3Rα; IL-3R-alpha; Interleukin-3 receptor subunit alpha |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.