missing translation for 'onlineSavingsMsg'
Learn More

CD123 Mouse anti-Human, FITC, Clone: 7G3, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 558663

 View more versions of this product

Product Code. 15899558

  • ONLINE EXCLUSIVE PRICE
    £424.00
    £406.65 / Each
    Save £17.35
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reacts with human CD123, the 70kDa IL-3 receptor α chain (IL-3Rα), which associates with the 120-140kDa β subunit. The β chain is shared with the receptors for interleukins IL-5 and GM-CSF. IL-3Rα is expressed on hematopoietic progenitors and plays an important role in hematopoietic progenitor cell growth and differentiation. This antibody has been reported to block the binding of 125I-IL-3 to high and low affinity IL-3 receptors. In functional experiments, this antibody was found to inhibit acute myeloid leukemia cell proliferation, basophil histamine release, endothelial cell-mediated IL-8 secretion, and neutrophil transmigration. This antibody has been reported to be useful for
immunoprecipitation, western blot and immunofluorescent staining for flow cytometry. At the Fifth HLDA Workshop, the human IL-3 receptor was designated CD123.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD123
Monoclonal
FITC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human IL-3Ra-transfected cells
100 Tests
Immunology
Human
IgG2a κ
Flow Cytometry
7G3
IL3RA; IL-3RA; IL-3Rα; IL-3R-alpha; Interleukin-3 receptor subunit alpha
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.