Learn More
CD123 Monoclonal Antibody (6H6), FITC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 11-1239-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The 6H6 monoclonal antibody reacts with human CD123, the alpha chain of the IL-3 receptor. This 60-70 kDa transmembrane protein binds to IL-3 with low affinity by itself, and when associated with CD131 (common beta chain) binds IL-3 with high affinity. CD123 is expressed by myeloid precursors, macrophages, dendritic cells, mast cells, basophils, and megakaryocytes. Applications Reported: This 6H6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 6H6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-3Ra (Interleukin-3 receptor subunit alpha) belongs to the type I cytokine receptor family and is a heterodimer with a unique alpha chain paired with the common beta (beta c or CDw131) subunit. IL-3Ra is responsible for the transmission of signal of Interleukin-3. IL-3Ra and IL-3Rb chain heterodimerization is induced by Interleukin-3, which is required for receptor activation but not high affinity binding. IL-3Ra is also reported that IL-3Ra subunits are overexpressed on acute myeloid leukemia (AML) blasts compared with normal hematopoietic cells and are thus potential targets for novel therapeutic agents.Specifications
CD123 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P26951 | |
IL3RA | |
Affinity chromatography | |
RUO | |
3563 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
6H6 | |
FITC | |
IL3RA | |
CD123, CDw123, SUT-1 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.