missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD123 (IL-3Rα) Mouse, anti-Human, APC, Clone: 6H6, STEMCELL Technologies
Mouse Monoclonal Antibody
£233.55 - £500.85
Specifications
| Antigen | CD123 (IL-3Rα) |
|---|---|
| Clone | 6H6 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17129091
|
Stemcell Technologies
60110AZ.1 |
25 Tests |
£233.55
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17119091
|
Stemcell Technologies
60110AZ |
100 Tests |
£500.85
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The 6H6 antibody reacts with human CD123 (IL-3 receptor subunit α), an ∼70 kDa type I transmembrane glycoprotein belonging to the type I cytokine receptor family (type 5 subfamily) and the immunoglobulin (Ig) superfamily
- CD123 constitutes the ligand-binding α chain of the heterodimeric IL-3 receptor
- CD123 binds IL-3 with low affinity per se, but when associated with CD131 (the signal-transducing β chain of the receptor), high-affinity binding of IL-3 is observed
- CD123 is expressed by hematopoietic progenitor cells, endothelial cells, basophils, eosinophils, mast cells, monocytes, macrophages, dendritic cells, megakaryocytes, a subset of B cells, and by neutrophils if cultured in the presence of granulocyte-macrophage colony-stimulating factor (GM-CSF)
- IL-3 binding to CD123 stimulates proliferation, differentiation, and viability of hematopoietic cells
- The 6H6 antibody does not inhibit binding of IL-3 to either CD123 or the IL-3 receptor
Specifications
| CD123 (IL-3Rα) | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| APC | |
| Rhesus Monkey, Sooty Mangabey, Human | |
| Primary |
| 6H6 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title