Learn More
CD122 Monoclonal Antibody (TU27), FITC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 11-1228-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: This TU27 monoclonal antibody reacts with human CD122, which is also known as interleukin-2 receptor beta (IL-2R beta). This receptor is expressed on NK cells, activated T cells, some B cells, monocytes, and T cell leukemias. CD122 can also be detected during early NK cell commitment. IL-2R beta associates with the IL-2R alpha (CD25) or IL-15R alpha and common gamma chain (CD132) to form the high affinity IL-2 and IL-15 receptors, respectively. By itself, this membrane receptor can bind IL-2, but with intermediate affinity. The binding of IL-2 results in internalization of the cytokine, tyrosine phosphorylation of IL-2Rbeta, and activation of signaling cascades such as the JAK/STAT pathway. Applications Reported: This TU27 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TU27 antibody has been pre-titrated and tested by flow cytometric analyis of normal human peripheral blood cells. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD122 (Interleukin 2 receptor) is involved in T cell-mediated immune responses, primarily expressed in the hematopoietic system and is present in 3 forms. The low affinity form of CD122 is a monomer of the alpha subunit and is not involved in signal transduction. The intermediate affinity form consists of an alpha/beta subunit heterodimer, while the high affinity form consists of an alpha/beta/gamma subunit heterotrimer. Both the intermediate and high affinity forms of the receptor are involved in receptor-mediated endocytosis and transduction of mitogenic signals from CD122. The protein encoded by the CD122 gene represents the beta subunit and is a type I membrane protein. Diseases associate with CD122 protein dysfunction include oligoarticular juvenile idiopathic arthritis and rheumatoid factor negative juvenile idiopathic arthritis.Specifications
CD122 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P14784 | |
Il2rb | |
Affinity chromatography | |
RUO | |
3560 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
TU27 | |
FITC | |
Il2rb | |
CD122; CD122 antigen; high affinity IL-2 receptor beta subunit; high affinity IL-2 receptor subunit beta; IL-15 receptor beta chain; IL15RB; IL15Rbeta; IL-15Rbeta; IL-2 receptor beta chain; IL-2 receptor subunit beta; IL-2/15 receptor-beta; Il-2/15Rbeta; IL-2R subunit beta; IL2RB; IL-2RB; IL2RBC; Il-2Rbeta; interleukin 15 receptor, beta; interleukin 2 receptor subunit beta; interleukin 2 receptor, beta; interleukin 2 receptor, beta chain; Interleukin-15 receptor subunit beta; Interleukin-2 receptor subunit beta; p70; p70-75; p75 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.