missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD122/IL2RB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13602-1-AP-150UL
This item is not returnable.
View return policy
Description
CD122 (Interleukin 2 receptor) is involved in T cell-mediated immune responses, primarily expressed in the hematopoietic system and is present in 3 forms. The low affinity form of CD122 is a monomer of the alpha subunit and is not involved in signal transduction. The intermediate affinity form consists of an alpha/beta subunit heterodimer, while the high affinity form consists of an alpha/beta/gamma subunit heterotrimer. Both the intermediate and high affinity forms of the receptor are involved in receptor-mediated endocytosis and transduction of mitogenic signals from CD122. The protein encoded by the CD122 gene represents the beta subunit and is a type I membrane protein. Diseases associate with CD122 protein dysfunction include oligoarticular juvenile idiopathic arthritis and rheumatoid factor negative juvenile idiopathic arthritis.Specifications
| CD122/IL2RB | |
| Polyclonal | |
| Unconjugated | |
| IL2RB | |
| CD122, IL 2 receptor subunit beta, IL 2R subunit beta, IL 2RB, IL2RB, interleukin 2 receptor, beta, P70 75, p75 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16185, 3560 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P14784, P16297 | |
| IL2RB | |
| CD122/IL2RB Fusion Protein Ag3929 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction