missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD122/CD132 Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen CD122/CD132 Biosimilar
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283665
GREENER_CHOICE
View Documents
Invitrogen™
MA558661
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30284605
View Documents
Invitrogen™
MA558662
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Singapore ASTR patent anti-IL-2R beta/IL-2R gamma .

CD122 and CD132 are integral subunits of the interleukin-2 receptor (IL-2R) complex, which also includes the alpha chain (CD25). They are respectively known as the beta chain (CD122) and the gamma chain (CD132). These genes are expressed in various cell types and are crucial for immunological responses. The heterodimeric CD122/CD132 receptor subunits are vital for the signal transduction of interleukins like IL-2 and IL-15, which are critical for T cell proliferation and activation. CD122, in particular, has an extensive intracellular region, making it indispensable for downstream signaling. Both CD122 and CD132 contribute to the receptor's function by binding IL-2 and other cytokines, facilitating their role in immune regulation and homeostasis. Studies suggest that CD122 and CD132 may be expressed by fibroblast-like synoviocytes (FLS) and dermal fibroblasts (DF), underscoring their broader biological significance beyond immune cells. This receptor complex is also a focal point in cancer immunotherapy strategies due to its role in modulating T regulatory cells (Tregs) and effector T cells.
Specifications

Specifications

CD122/CD132 Biosimilar
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG1
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.