Learn More
CD120b Rat anti-Human, Brilliant Violet 421, Clone: hTNFR-M1, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 742984
Description
The hTNFR-M1 antibody specifically recognizes the extracellular domain of the 75 kDa transmembrane receptor for the human cytokines, tumor necrosis factor (TNF or TNF-α) and lymphotoxin-alpha (LT-α3, aka, lymphotoxin or TNF-β). This receptor is referred to as the p75 or Type II Tumor Necrosis Factor Receptor (TNFRII) [aka, CD120b]. Human TNFRII proteins are expressed by hematopoietic cells including macrophages, neutrophils, lymphocytes, thymocytes and mast cells. TNFRII is expressed by a variety of other cell types including endothelial cells, cardiac myocytes and prostate cells. Naive B cells express very low or undetectable levels of TNFRII whereas mature erythrocytes and platelets are uniformly negative for TNFRII expression. The immunogen used to generate the hTNFR-M1 hybridoma was COS- expressed recombinant human TNFRII.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications
| CD120b | |
| hTNFR-M1 | |
| Brilliant Violet 421 | |
| TNFRSF1B, p75 TNFR, p75 TNF Receptor, TNFR80, TNF-RII, TNF-R2 | |
| COS-expressed recombinant human TNFRII | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.