missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD11d Monoclonal Antibody (CA11.8H2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528277
This item is not returnable.
View return policy
Description
Mouse anti Dog CD11d antibody, clone CA11.8H2 recognizes the canine CD11d cell surface antigen, also known as integrin alpha d subunit.
Integrin alpha-D/beta-2 is a receptor for ICAM3 and VCAM1. May play a role in the atherosclerotic process such as clearing lipoproteins from plaques and in phagocytosis of blood-borne pathogens, particulate matter, and senescent erythrocytes from the blood.
Specifications
| CD11d | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| 0 | |
| ITGAD | |
| Affinity purified CD11 antigen from canine spleen. | |
| 100 μg | |
| Primary | |
| Canine | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| CA11.8H2 | |
| Unconjugated | |
| ITGAD | |
| ADB2; beta-2 integrin alphaD subunit; CD11 antigen-like family member D; CD11d; FLJ39841; integrin alpha X (Cd11c); integrin alpha-D; integrin subunit alpha D; integrin, alpha D; ITGAD; Itgax; Leukointegrin alpha D | |
| Mouse | |
| Protein A | |
| RUO | |
| 489930 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction