missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11d/Integrin alpha D Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55162-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to CD11d.
Integrin alpha-D/beta-2 is a receptor for ICAM3 and VCAM1. May play a role in the atherosclerotic process such as clearing lipoproteins from plaques and in phagocytosis of blood-borne pathogens, particulate matter, and senescent erythrocytes from the blood.Specifications
| CD11d/Integrin alpha D | |
| Polyclonal | |
| Unconjugated | |
| ITGAD | |
| ADB2, CD11d, Integrin alpha D, integrin, alpha D, ITGAD, Leukointegrin alpha D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3681 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13349 | |
| ITGAD | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction