missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Mouse anti-Human, eFluor 570, Clone: 118/A5, eBioscience™
Mouse Monoclonal Antibody
Brand: Invitrogen 41-9761-80
This item is not returnable.
View return policy
Description
The monoclonal antibody 118/A5 recognizes human CD11c, a 150 kD type I transmembrane protein which is also known as integrin alpha X. Integrins are heterodimeric integral membrane proteins made up of an alpha and a beta chain and are involved in cell adhesion. The CD11/CD18 adhesion molecules consist of three heterodimers sharing a common beta subunit (CD18) with a distinct alpha subunit (CD11a, CD11b, CD11c). The CD11c/CD18 complex is expressed on dendritic cells (highly expressed on type 1 myeloid DCs), monocytes, macrophages, granulocytes, NK cells, and subsets of T and B cells. CD11c is also expressed in hairy cell leukemias, acute nonlymphocytic leukemias, and some B-cell chronic lymphocytic leukemias. CD11c plays a role in adhesion and CTL killing through its interactions with CD54, iC3b and fibrinogen.
Specifications
| CD11c | |
| Monoclonal | |
| 0.2mg/mL | |
| aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
| Mouse | |
| RUO | |
| Human | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Frozen), Immunomicroscopy | |
| 118/A5 | |
| eFluor 570 | |
| Integrin alpha X | |
| 25 μg | |
| Primary | |
| Store at 2-8°C. Do not freeze. Light-sensitive material. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction