missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human CD11c Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
£170.00 - £486.00
Specifikationer
| Antigen | CD11c |
|---|---|
| Clone | ICRF 3.9 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation, CyTOF-ready |
| Applications | Flow Cytometry, Immunoprecipitation, CyTOF |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
18784057
![]() |
R&D Systems
MAB1777-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18159500
![]() |
R&D Systems
MAB1777 |
500 μg |
£486.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
CD11c Monoclonal specifically detects CD11c in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF-ready.Especificaciones
| CD11c | |
| LYOPH | |
| Flow Cytometry, Immunoprecipitation, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, Integrin alpha X, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), ITGAX, Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, p150,95 alpha, SLEB6 | |
| CD11c | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| ICRF 3.9 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3687 | |
| Rheumatoid synovial fluid cells and human monocyte-derived fibronectin | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD11c. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto