Learn More
CD11c Mouse anti-Human, BUV395, Clone: S-HCL-3, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 744440
Description
The S-HCL-3 monoclonal antibody specifically binds to CD11c which is encoded by ITGAX (Integrin alpha X, complement component 3 receptor 4 subunit). CD11c is a ñ150 kDa type I transmembrane glycoprotein that is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. CD11c is also present on hairy-cell leukemias and on some acute myeloid leukemias (AML). The S-HCL-3 antibody stains sinus histiocytes of lymph node, tonsil, and spleen; macrophages in the cortex and medulla of thymus; Kupffer cells in liver; elongated intertubular cells in kidney; and alveolar macrophages in lung tissue. In malignant tissue, the antibody stains true histiocytic lymphomas; it does not stain carcinomas or T or B lymphomas. CD11c associates with CD18 (ITGB2/Integrin beta-2) to form the CD11c/CD18 heterodimer which is likewise known as Leukocyte adhesion receptor p150,95 or Complement receptor type 4 (CR4). Ligands for this heterodimeric adhesion receptor include fibrinogen, iC3b, CD23, and CD54 (ICAM-1).
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Specifications
| CD11c | |
| S-HCL-3 | |
| Brilliant Ultraviolet 395 | |
| ITGAX, Integrin alpha-X, AlphaX integrin chain, CR4, Leu M5, p150/95 alpha | |
| Human Hairy-Cell Leukemia Cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.