Learn More
CD11c Mouse anti-Human, Brilliant Ultraviolet 661, Clone: B-ly6, BD Horizon™
Mouse Monoclonal Antibody
£188.32 - £430.16
Specifications
| Antigen | CD11c |
|---|---|
| Clone | B-ly6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Ultraviolet 661 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17238014
|
BD Biosciences
612968 |
25 Tests |
£197.00 £188.32 / 25 tests Save £8.68 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17228014
|
BD Biosciences
612967 |
100 Tests |
£445.00 £430.16 / 100 tests Save £14.84 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The B-ly6 monoclonal antibody specifically binds to the 150 kDa adhesion glycoprotein CD11c (p150, integrin α chain). CD11c is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. It associates with CD18 to form the CD11c/CD18 complex that binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 plays a role as an adhesion molecule that mediates cellular binding to ligands expressed on stimulated epithelium and endothelium.
The antibody was conjugated to BD Horizon BUV661 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is a tandem fluorochrome of BD Horizon BUV395 with an Ex Max of 348-nm and an acceptor dye with an Em Max at 661-nm. BD Horizon Brilliant BUV661 can be excited by the ultraviolet laser (355 nm) and detected with a 670/25 filter and a 630nm LP. Due to cross laser excitation of this dye, there may be significant spillover into channels detecting APC-like emissions (eg, 670/30-nm filter).
Specifications
| CD11c | |
| Flow Cytometry | |
| Brilliant Ultraviolet 661 | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| B-ly6 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Spleen Cells from Human with Hairy Cell Leukemia | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.