Learn More
CD11c Monoclonal Antibody (N418), PE-Cyanine5.5, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 35-0114-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The N418 monoclonal antibody reacts with mouse CD11c, the integrin alpha X. CD11c non-covalently associates with beta 2 integrin to form the CD11c/CD18 heterodimer. CD11c is expressed by dendritic cells, a subset of Intestinal Intraepithelial Lymphocytes (IEL) and some activated T cells. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions. N418 binds to CD11c on splenic dendritic cells in the T-dependent areas of mouse spleen and precipitates a 150, 90 kDa heterodimer. Applications Reported: This N418 antibody has been reported for use in flow cytometric analysis. Applications Tested: This N418 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD11, along with CD18, form a heterodimer adhesion molecule. In particular, CD11 is composed of CD11a, CD11b and CD11c. CD11a is a leukocyte marker that is expressed in B and T lymphocytes, macrophages, monocytes, neutrophils, basophils and eosinophils. CD11b is primarily expressed by monocytes, macrophages, natural killer cells, some B and T-cells, and granulocytes. CD11c is expressed in monocytes, macrophages, natural killer cells, some granulocytes and less so in a subset of lymphocytes.Specifications
CD11c | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9QXH4 | |
Itgax | |
Affinity chromatography | |
RUO | |
16411 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
N418 | |
PE-Cyanine5.5 | |
Itgax | |
AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6 | |
Armenian Hamster | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
For Research Use Only.