Learn More
CD11c Hamster, anti-Mouse, PerCP-Cyanine5.5, Clone: N418, STEMCELL Technologies
Hamster Monoclonal Antibody
Brand:  Stemcell Technologies 60002PS.1
Encompass_Preferred
 View more versions of this product
Product Code. 17188331
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • The N418 antibody reacts with CD11c (αX integrin), a 150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD18 (β2 integrin) to form a heterodimeric cell surface adhesion receptor
  • Through its interaction with ligands such as iC3b, fibrinogen, and CD54, the CD11c/CD18 receptor is involved in several immune response processes, including cell migration, stimulation of cytokine production by monocytes and macrophages, T cell proliferation, leukocyte recruitment, and phagocytosis
  • In mice, CD11c is expressed on dendritic cells, macrophages, monocytes, granulocytes, NK cells, and a subset of T cells
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD11c Positive Selection Kit II (Catalog #18780)
Specifications
Specifications
CD11c
Monoclonal
PerCP-Cyanine5.5
25 μg
Mouse
Dendritic Cells
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
N418
Hamster