Learn More
CD11c Hamster, anti-Mouse, PE, Clone: N418, STEMCELL Technologies
Hamster Monoclonal Antibody
Brand: Stemcell Technologies 100-0445
This item is not returnable.
View return policy
Description
- The N418 antibody reacts with CD11c (αX integrin), a 150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD18 (β2 integrin) to form a heterodimeric cell surface adhesion receptor
- Through its interaction with ligands such as iC3b, fibrinogen, and CD54, the CD11c/CD18 receptor is involved in several immune response processes, including cell migration, stimulation of cytokine production by monocytes and macrophages, T cell proliferation, leukocyte recruitment, and phagocytosis
- In mice, CD11c is expressed on dendritic cells, macrophages, monocytes, granulocytes, NK cells, and a subset of T cells
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD11c Positive Selection Kit II (Catalog #18780)
Specifications
| CD11c | |
| Monoclonal | |
| PE | |
| 25 μg | |
| Mouse |
| CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| N418 | |
| Hamster | |
| Primary | |
| Dendritic Cells |