missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Hamster, anti-Mouse, APC, Clone: N418, STEMCELL Technologies
Hamster Monoclonal Antibody
£137.70 - £436.05
Specifications
| Antigen | CD11c |
|---|---|
| Clone | N418 |
| Applications | CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17128331
|
Stemcell Technologies
60002AZ.1 |
25 μg |
£137.70
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17118331
|
Stemcell Technologies
60002AZ |
100 μg |
£436.05
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The N418 antibody reacts with CD11c (αX integrin), a 150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD18 (β2 integrin) to form a heterodimeric cell surface adhesion receptor
- Through its interaction with ligands such as iC3b, fibrinogen, and CD54, the CD11c/CD18 receptor is involved in several immune response processes, including cell migration, stimulation of cytokine production by monocytes and macrophages, T cell proliferation, leukocyte recruitment, and phagocytosis
- In mice, CD11c is expressed on dendritic cells, macrophages, monocytes, granulocytes, NK cells, and a subset of T cells
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD11c Positive Selection Kit II (Catalog #18780)
Specifications
| CD11c | |
| CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| APC | |
| Mouse | |
| Primary |
| N418 | |
| Monoclonal | |
| Hamster | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title