missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Antibody (ITGAX/1243), Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £375.00
Specifications
| Antigen | CD11c |
|---|---|
| Clone | ITGAX/1243 |
| Classification | Monoclonal |
| Form | Purified |
| Isotype | IgG κ |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18760084
|
Novus Biologicals
NBP2-44599-0.02mg |
£221.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18108747
|
Novus Biologicals
NBP2-44599-0.1MG |
0.1 mg |
£375.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
CD11c Monoclonal specifically detects CD11c in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| CD11c | |
| Monoclonal | |
| IgG κ |
| ITGAX/1243 | |
| Purified | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title