missing translation for 'onlineSavingsMsg'
Learn More

CD11c Antibody (ITGAX/1243), DyLight 680, Novus Biologicals™

Product Code. 18421254 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421254 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421254 Supplier Novus Biologicals Supplier No. NBP254432FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD11c Monoclonal antibody specifically detects CD11c in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone ITGAX/1243
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6
Host Species Mouse
Immunogen Recombinant fragment (aa 637-827) of human CD11c protein (exact sequence is proprietary) (Uniprot: P20702)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Dendritic Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3687
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.