missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Antibody (BU15), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-45018MFV610
This item is not returnable.
View return policy
Description
CD11c Monoclonal antibody specifically detects CD11c in Human, Monkey samples. It is validated for Flow Cytometry
Specifications
| CD11c | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
| Flow Cytometry | |
| BU15 | |
| Flow Cytometry | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
| Dendritic cells of synovial fluid | |
| 0.1 mL | |
| Dendritic Cell Markers, Signal Transduction | |
| 3687 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction