Learn More
CD11b Rat anti-Mouse, CoraLite 594, Clone: M1/70, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech CL594-65055-100TESTS
Description
The CD11b antibody from Proteintech is a rat monoclonal antibody generated with c57bl/10 mouse splenic t cells and concanavalin a-activated c57bl/10 splenocytes of mouse CD11b. This antibody recognizes mouse antigen. The CD11b antibody has been validated for the following applications: FC analysis. This antibody is coralite™594.
Specifications
| CD11b | |
| Monoclonal | |
| 5 μl/Test | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| M1/70 | |
| CoraLite 594 | |
| 100 Tests | |
| 16409 | |
| IgG2b |