Learn More
CD11b Mouse, anti-Human, FITC, Clone: ICRF44, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60040FI.1
This item is not returnable.
View return policy
Description
- The ICRF44 antibody reacts with an extracellular epitope on CD11b (integrin αM), an ∼170 kDa type 1 transmembrane glycoprotein which associates non-covalently with CD18 to form the heterodimeric Mac-1 receptor
- Through its interactions with ligands such as ICAM-1 (CD54), ICAM-2 (CD102), ICAM-4 (CD242), iC3b, heparin, and fibrinogen, Mac-1 influences several processes, including the adherence of neutrophils and monocytes to stimulated endothelium, and phagocytosis of complement-coated particles
- CD11b is expressed on the surface of granulocytes, monocytes, NK cells, dendritic cells, tissue macrophages, and subsets of T and B cells, and has been used as a marker to distinguish naive and memory CD8+ T cells
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human Monocyte Enrichment Kit (Catalog #19059), and for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411)
Specifications
| CD11b | |
| Monoclonal | |
| FITC | |
| 25 Tests | |
| Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human | |
| Myeloid Cells |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| ICRF44 | |
| Mouse | |
| Primary | |
| IgG1 κ |