missing translation for 'onlineSavingsMsg'
Learn More

CD11b/Mac-1 Mouse anti-Human, PE-Cy7, Clone: ICRF44, BD

Mouse Monoclonal Antibody

£56.57 - £358.08

Specifications

Antigen CD11b
Clone ICRF44
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE-Cyanine7
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15801239
View Documents
BD Biosciences
561685
25 Tests
ONLINE EXCLUSIVE PRICE
£58.85
£56.57 / Each
Save £2.28
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15829028
View Documents
BD Biosciences
557743
100 Tests
ONLINE EXCLUSIVE PRICE
£363.00
£358.08 / Each
Save £4.92
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The ICRF44 monoclonal antibody specifically binds to CD11b, the 165-kDa adhesion glycoprotein that associates with the 95-kDa integrin β2 (CD18) to form the CD11b/CD18 complex, also known as Mac-1 or CR3. CD11b is expressed on activated lymphocytes, monocytes, granulocytes, and a subset of NK cells. CD11b functions in cell-cell and cell-substrate interactions and is a receptor for iC3b, CD54 (ICAM-1), CD102 (ICAM-2) and CD50 (ICAM-3). This antibody significantly inhibits polymorphonuclear leukocyte aggregation in response to fMLP.

Specifications

Specifications

CD11b
Flow Cytometry
PE-Cyanine7
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
Mac-1α, integrin αM subunit, CR3 α chain
ICRF44
Monoclonal
Mouse
Human
Human monocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.